Learn More
Cayman Chemical Cholesteryl HomoLInolenate 1mg
SDP
Supplier:  Cayman Chemical 264691 MG
Encompass_Preferred
A cholesterol ester; accumulates in the adrenal gland of rabbits fed a diet high in linolenic acid; levels are decreased and positively correlate with cognitive decline in HIV-infected humans
Catalog No. 50-293-4364