Learn More
Cayman Chemical Cholesteryl Oleate 500mg
SDP
Supplier:  Cayman Chemical 26472500 MG
Encompass_Preferred
A cholesterol ester; has been used in the generation of nanoparticles and liposomes for drug delivery
Catalog No. 50-293-4358