Learn More
Cayman Chemical Cholesteryl PalmItate-d9 1mg
SDP
Supplier:  Cayman Chemical 281231 MG
Encompass_Preferred
An internal standard for the quantification of cholesteryl palmitate by GC- or LC-MS
Catalog No. 50-293-7735