Learn More
Medchemexpress LLC Cholestyramine | 11041-12-6 | 435.1 g/mol | C27H47ClN2 | 10 G
SDP
Supplier:  Medchemexpress LLC HY10408110G
Encompass_Preferred
Cholestyramine is a chloride-form anion-exchange bile acid binding resin supplied as a white to off-white powder for research and analytical use. It sequesters intestinal bile acids, reducing bile acid absorption and driving increased bile acid synthesis from cholesterol, and is used in biochemical studies and in vitro bile acid removal.
  • Binds intestinal bile acids effectively.
  • Supplied as a powder for easy suspension and handling.
  • Contains specified chloride content and exchange capacity for predictable performance.
  • Certificate of analysis reports chloride content, loss on drying, and exchange capacity.
  • Suitable for biochemical and physiological research involving bile acid sequestration.
Catalog No. 50-002-41748