Learn More
Medchemexpress LLC Cholestyramine 50g | 11041-12-6 | 50G
SDP
Supplier:  Medchemexpress LLC HY10408150G
Encompass_Preferred
Cholestyramine (Colestyramine) is a bile acid binding resin and can inhibit intestinal bile acid absorption which results in the increasing bile acid synthesis from cholesterol
Catalog No. 50-002-42208