Learn More
Medchemexpress LLC Cholestyramine resin | 11041-12-6 | 13.0-17.0% | 435.1 g/mol | C27H47N | 100 G
SDP
Supplier:  Medchemexpress LLC HY104081100G
Encompass_Preferred
Cholestyramine is a bile acid-binding resin used in research to sequester intestinal bile acids, reducing enterohepatic recirculation and increasing bile acid synthesis from cholesterol. Supplied as a white to off-white powder, it is intended for laboratory formulations and in vivo suspension studies.
  • Binds intestinal bile acids, reducing bile acid reabsorption.
  • Increases bile acid synthesis from cholesterol.
  • Powder form, white to off-white, suitable for suspension formulations.
  • Low solubility in water and common solvents (insoluble or slightly soluble).
  • Can be formulated as a suspension in 0.5% CMC-Na for in vivo dosing (approximately 60 mg/mL).
  • Stable when stored as a powder at -20°C (3 years) or 4°C (2 years).
Catalog No. 50-002-42568