Learn More
Medchemexpress LLC Cholestyramine resin | 11041-12-6 | MFCD00130784 | 16.1% | Polymeric anion-exchange resin; no defined molecular formula | 5 G
SDP
Supplier:  Medchemexpress LLC HY1040815G
Encompass_Preferred
Cholestyramine (colestyramine) is a crosslinked chloride-form anion-exchange resin supplied as a powder. It binds bile acids in the intestine, reducing enterohepatic bile acid recycling and promoting increased bile acid synthesis from cholesterol. It is used in biochemical research, formulation studies, and assays that require bile acid sequestration.
  • Binds bile acids in the intestine, reducing enterohepatic recycling.
  • Promotes increased bile acid synthesis from cholesterol.
  • Supplied as a powdered, polymeric anion-exchange resin suitable for research and formulation.
  • Typical chloride assay 13-17% (example 16.1% reported on COA).
  • No single defined molecular weight because the product is a crosslinked polymer.
Catalog No. 50-002-42524