Learn More
Medchemexpress LLC Cholic acid 7-sulfate | 60320-05-0 | 99.9% | 488.63 g/mol | C24H40O8S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1268555MG
Encompass_Preferred
Cholic acid 7-sulfate is a sulfated bile acid used as a research compound. It functions as a potent TGR5 (GPBAR1) agonist and an endogenous ligand for MR1, and is used to study bile acid signaling, GLP-1 secretion, glucose tolerance, and MAIT cell biology in metabolic and immunology research.
  • High reported purity (99.92%).
  • Potent TGR5 agonist (EC50 = 0.17 μM).
  • Acts as an MR1 ligand affecting MAIT cell survival and function.
  • Suitable for in vitro and in vivo research applications.
  • Offered in small research pack sizes for preclinical studies.
Catalog No. 50-002-64586