Learn More
Medchemexpress LLC Cholic acid-biotin | 95.0% | 809.11 | C42H72N4O9S | 10 MG

Supplier: Medchemexpress LLC HYN0324F10MG
Cholic acid-biotin is a biotin-labeled derivative of cholic acid supplied for research use as a solid reagent. It is used in biochemical and binding assays, facilitates fat absorption and cholesterol excretion, and is reported to have oral activity. The compound is provided in small milligram pack sizes for laboratory experiments.
- Biotin-labeled cholic acid suitable for biochemical assays
- Purity 95.0%
- White to off-white solid; DMSO soluble (100 mg/mL)
- Molecular weight 809.11; formula C42H72N4O9S
- Store solid at room temperature protected from light; in solution store at -80°C up to 6 months
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.