Learn More
Medchemexpress LLC Cholic acid-d4 | 116380-66-6 | 99.9% | 412.60 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0324S50MG
Encompass_Preferred
Cholic acid-d4 is the deuterium-labeled Cholic acid. Cholic acid is a major primary bile acid produced in the liver and usually conjugated with glycine or taurine. It facilitates fat absorption and cholesterol excretion. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Major primary bile acid.
  • Produced in the liver.
  • Usually conjugated with glycine or taurine.
  • Facilitates fat absorption.
  • Facilitates cholesterol excretion.
Catalog No. 50-004-27306