Learn More
Cayman Chemical CholIc AcId nIlIde 5mg
SDP
Supplier:  Cayman Chemical 261445 MG
Encompass_Preferred
A synthetic bile acid and derivative of cholic acid; inhibits the germination of C. difficile strain R20291 spores in vitro (IC50 = 1.8 μM)
Catalog No. 50-293-6261