Learn More
AG Scientific Inc Choline Chloride 2.5 KG
Supplier:  AG Scientific Inc C27612.5KG
Encompass_Preferred
Choline chloride is used in choline release assays. Acts as endogenous agonist of sigma-1 receptors. Standard to analyze relationships between methionine and choline metabolism.Choline chloride is also used as an additive in poultry feed to accelerate growth in chickens and as an additive for hydraulic fracturing fluids
Catalog No. 50-226-8275