Learn More
Alkali Scientific Choline Chloride, 250 Grams
SDP
Supplier:  Alkali Scientific CHO004CP
Encompass
Choline Chloride, a quaternary ammonium compound, is commonly used in laboratory settings as a vital component in molecular biology and biochemistry. This 250g powder form of choline chloride is essential for studying lipid metabolism, enzyme activity, and as a nutrient supplement in cell culture media. It is used extensively in the production of phosphatidylcholine, a key phospholipid in cellular membranes. Additionally, choline chloride plays an important role in understanding neurobiology, liver function, and metabolic disorders. This reagent also finds applications in agricultural research, particularly in animal feed and growth studies.
Catalog No. 50-006-60830