Learn More
Apexbio Technology LLC Choline Chloride 67-48-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B17035.1
Encompass_Preferred
Choline chloride (CAS 67-48-1) is a quaternary ammonium compound that serves as a precursor of the neurotransmitter acetylcholine and is integral to phospholipid biosynthesis It functions as a methyl group donor in various metabolic pathways notably in the synthesis of phosphatidylcholine and betaine In biomedical research choline chloride is utilized to investigate cellular membrane composition lipid metabolism and neurological functions It also finds application in studies of nutritional requirements and epigenetic regulation as well as in cell culture media to support cellular proliferation and viability
Catalog No. 50-000-94222