Learn More
Medchemexpress LLC Choline-d4 (chloride) | 285979-70-6 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1337S50MG
Encompass_Preferred
Choline-d4 (chloride) is the deuterium labeled Choline chloride. Choline chloride is an organic compound and a quaternary ammonium salt, an acyl group acceptor and choline acetyltransferase substrate, and also an important additive in feed, especially for chickens where it accelerates growth. It is for research use only, and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12102