Learn More
Cayman Chemical Cholyl-putrescIn 5mg
SDP
Supplier:  Cayman Chemical 408995 MG
Encompass_Preferred
A polyamine bile amidate; has been found in lion and mouse, as well as serval, cheetah, and tiger fecal samples; has been used as a standard for the detection of endogenous bile acid amidates in mammalian intestinal and fecal samples
Catalog No. 50-294-4825