Learn More
Selleck Chemical LLC Chondroitin sulfate S2416-25mg
SDP
Supplier:  Selleck Chemical LLC S241625MG
Encompass_Preferred
Chondroitin sulfate (CS) is a major component of the extracellular matrix (ECM) of many connective tissues including cartilage bone skin ligaments and tendons It reduces the IL-1 -induced nuclear factor-kB (NF- B) translocation in chondrocytes in vitro
Catalog No. 50-311-2141