Learn More
Amsbio LLC Chondroitinase ABC protease free (lyophilized) 10U/vial
Supplier:  Amsbio LLC AMS.E102810
Encompass
Chondroitinase ABC is purified from Proteus vulgaris by cation exchange chromatography. It catalyzes thedegradation of chondroitin sulfate A, chondroitin sulfate C, dermatan sulfate, chondroitin and hyaluronan to mainly disaccharideswith D-4hexuronate by the eliminative cleavage of 1,4-B-hexosaminyl linkages. It does not act on keratan sulfate, heparin and heparan sulfate.
Catalog No. 50-204-1244