Learn More
CPC Scientific H-Thr-Cys-Asp-Asp-Pro-Arg-Phe-Gln-Asp-Ser-Ser-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific HCG001B
Encompass
This highly immunogenic undecapeptide generates hCGspecific antibodies ONE-LETTER SEQUENCE: TCDDPRFQDSSMOLECULAR FORMULA: C50H76N16O21S1MOLECULAR WEIGHT:1269.32STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [165174-59-4], RESEARCH AREA: ImmunomodulationREFERENCES: K.S.N.Iyer et al., Int. J. Pept. Protein Res., 39, 137 (1992)
Catalog No. 50-210-9415