Learn More
Genetex Inc Chromogranin A Monoclonal Antibody [MSVA-380R] HistoMAX™ (GTX04370) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04370
Encompass_Preferred
The Chromogranin A antibody [MSVA-380R] HistoMAX™ (GTX04370) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human pancreas tissue using GTX04370 Chromogranin A antibody [MSVA-380R] HistoMAX. Islet cells of the pancreas show intense chromogranin A staining.
Catalog No. NC3664116