Learn More
Chemglass Life Sciences Column, Chromatography, 35/20 Socket Joint, 1in ID X 12in E.L., 2mm Stpk
SDP
Supplier:  Chemglass Life Sciences CG119920
Encompass_Preferred
Column, Chromatography, 35/20 Socket Joint, 1in ID X 12in E.L., 2mm Stopcock. Similar to CG-1198 but with a COARSE porosity disc sealed into the lower end of the column for packing support. Column is constructed using MEDIUM WALL tubing. Can be used with optional CG-1201 flow controller and CG-1200 reservoir. Lower end of the column has a round bottom. Connections between the column, reservoir and flow control adapter are spherical joints connected with CG-150 stainless steel pinchclamps.
Catalog No. CG119920