Learn More
Chemglass Life Sciences Column, Chromatography, 1 1/2in ID X 18in E.L., 2mm Stpk
SDP
Supplier:  Chemglass Life Sciences CG118716
Encompass_Preferred
Column, Chromatography, 1 1/2in ID X 18in E.L., 2mm Stopcock. Similar to CG-1186 but with a sealed in COARSE porosity fritted disc to support column packing. Lower end of the column has a round bottom. Column is constructed using MEDIUM WALL TUBING.
Catalog No. CG118716