Learn More
Drg International Inc CHROMagar™ VRE, dehydrated culture media, 5000 mL reconstituted
Supplier:  Drg International Inc VR952
Encompass
CHROMagar™chromogenic plates are easy to prepare - just add water to the dehydrated media, bring to a boil, let it cool down and pour! Use the entire pack, or if there is a need for a smaller number of plates, just a portion. CHROMagar™ products are sold in dehydrated form. DRG offers the 5L size (after reconstitution). If kept under appropriate storage temperatures, CHROMagar™ powder products have a long shelf life (2-3 years). This flexibility is essential to avoid the costly waste resulting from unused plates. This product requires a refrigerated supplement to use properly; these are shipped free of charge along with the product. Supplements contain compounds that cannot be heated, therefore the compounds would degrade in the base if stored together for a long time.
Catalog No. NC0838667