Learn More
Medchemexpress LLC Chroman 1 dihydrochloride | 1273579-40-0 | MFCD23098792 | 99.1% | 509.43 | C24H30Cl2N4O4 | 10 MG

Supplier: Medchemexpress LLC HY15392A10MG
Chroman 1 dihydrochloride is a highly potent and selective Rho-associated protein kinase (ROCK) inhibitor, with markedly greater activity against ROCK2 than ROCK1. Supplied as a dihydrochloride solid salt with high purity and good solubility, it is used in biochemical and cellular studies to probe ROCK signaling pathways.
- Highly potent, selective ROCK2 inhibitor (IC50 1 pM).
- Also inhibits ROCK1 and MRCK (IC50 52 pM and 150 nM).
- High purity: 99.11%.
- Soluble in water and DMSO: 100 mg/mL.
- Supplied as a solid dihydrochloride salt for stability.
- Recommended storage: sealed and away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.