Learn More
Medchemexpress LLC Chroman 1 dihydrochloride | C24H30Cl2N4O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY15392A5MG
Encompass_Preferred
Chroman 1 dihydrochloride is a highly potent and selective ROCK inhibitor, showing more potency against ROCK2 (IC50=1 pM) than ROCK1 (IC50=52 pM). It also exhibits inhibitory activity against MRCK, with an IC50 of 150 nM. In vitro studies indicate that Chroman 1 (50 nM, 24 h) inhibits caspase-3/7 activation and reduces apoptosis in human pluripotent stem cells.
  • Highly potent and selective ROCK inhibitor
  • More potent against ROCK2 (IC50=1 pM) than ROCK1 (IC50=52 pM)
  • Inhibits MRCK (IC50=150 nM)
  • Inhibits caspase-3/7 activation
  • Reduces apoptosis in human pluripotent stem cells
Catalog No. 50-003-76329