Learn More
Hach Company ChromaVer 3 Chromium Reagent Powder Pillows, 25 mL, pk/100
Supplier:  Hach Company 1206699
Encompass
For hexavalent chromium determination by the Diphenylcarbohydrazide Method. USEPA accepted for reporting for wastewater analyses using Hach Method 8023. Range: up to 0.70 mg/L Cr. Sample Size: 25 mL. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-4853