Learn More
Bellco Glass, Inc. ChromCap 100 HPLC Cap Assembly , 38-430, Adapter w Vent Hole, Two (2x) Ports- OD Tubing 3.2mm (1/8") or 1.6mm (1/16"), 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800069
Encompass_Preferred
PUREGRIP HPLC Solvent Reservoir used in upstream Caps & Bottle system offer the safest & most ergonomic lab safety solution in the industry. This system come with patented VersaCap , a Class VI PTFE Manifold, chemically resistant Viton O-Ring.The sealed VersaCap design that offers eliminates tubes twisting as you thread the cap & eliminates the tubes from sliding in & out of the container. Each assembly comes with 38-430 Cap & Manifold Assy., High quality fittings for 2 x 3.2mm(1/8″) OD Tubing, 2 x 1.6mm(1/16″) OD Tubing & 2 x Plugs of 6.4mm(1/4″) for blocking of unused ports.
Catalog No. 50-255-0693