Learn More
NANOMICRO TECHNOLOGIES INC ChromCore 120 C8 LC Analytical Column, 3 μm, 4.6 x 50 mm, Stainless Steel
Supplier:  NANOMICRO TECHNOLOGIES INC A00703001204605S
Encompass_Preferred
ChromCore 120 C8 columns are based on monodispersed high-purity porous silica particles densely bonded with C8 ligand and exhaustive end-capping to minimize undesired silanol activity designed for separating analytes with intermediate to high hydrophobicity
Catalog No. 50-003-32588