Learn More
NANOMICRO TECHNOLOGIES INC ChromCore 300 C8 LC Analytical Column, 3 μm, 2.1 x 150 mm, Stainless Steel
Supplier:  NANOMICRO TECHNOLOGIES INC A00703003002115S
Encompass_Preferred
ChromCore 300 C8 columns are based on monodispersed 300 pore high-purity spherical silica particles modified with C8 ligand and exhaustive end-capping They are designed for separating analytes of large size such as ginsenosides PEG derivatives peptides and small proteins
Catalog No. 50-003-32562