Learn More
NANOMICRO TECHNOLOGIES INC ChromCore AQ C18 LC Analytical Column, 3 μm, 3.0 x 150 mm, Stainless Steel
Supplier:  NANOMICRO TECHNOLOGIES INC A20103001803015S
Encompass_Preferred
ChromCore AQ C18 columns are based on proprietary C18 modified monodispersed silica particles for excellent aqueous compatibility They are the column of choice for applications requiring highly aqueous mobile phase and/or C18 selectivity
Catalog No. 50-003-32516