Learn More
NANOMICRO TECHNOLOGIES INC ChromCore Biphenyl LC Analytical Column, 3 μm, 3.0 x 100 mm, Stainless Steel
Supplier:  NANOMICRO TECHNOLOGIES INC A21103001203010S
Encompass_Preferred
ChromCore Biphenyl columns are based on monodispersed high-purity porous silica particles bonded with biphenyl ligand and exhaustive end-capping They are designed for applications that require different selectivity towards aromatic analytes compared to ChromCore Phenyl or ChromCore PFP
Catalog No. 50-003-32532