Learn More
NANOMICRO TECHNOLOGIES INC ChromCore C30 Plus LC Analytical Column, 5 μm, 4.6 x 250 mm, Stainless Steel
Supplier:  NANOMICRO TECHNOLOGIES INC A06205002004625S
Encompass_Preferred
ChromCore C30 Plus columns are based on monodispersed high-purity porous silica particles bonded with C30 ligand and exhaustive end-capping designed for separating structurally related compounds with large molecular size
Catalog No. 50-003-32599