Learn More
NANOMICRO TECHNOLOGIES INC ChromCore HILIC-Amide LC Analytical Column, 3 μm, 4.6 x 150 mm, Stainless Steel
Supplier:  NANOMICRO TECHNOLOGIES INC A06803001204615S
Encompass_Preferred
ChromCore HILIC-Amide columns are based on proprietary amido modified monodispersed high-purity silica particles They provide high polarity and are designed for separating a broad range of polar compounds
Catalog No. 50-003-32534