Learn More
NANOMICRO TECHNOLOGIES INC ChromCore HILIC-ZW LC Analytical Column, 5 μm, 4.6 x 250 mm, Stainless Steel
Supplier:  NANOMICRO TECHNOLOGIES INC A11405001804625S
Encompass_Preferred
ChromCore HILIC-ZW columns are based on innovative monodispersed particle technology and unique zwitterionic sulfobetaine column chemistry They provide salt exchange property and high polarity featuring a selectivity complementary to other HILIC columns for separating highly polar compounds
Catalog No. 50-003-32527