Learn More
NANOMICRO TECHNOLOGIES INC ChromCore Polar C18 LC Analytical Column, 3 μm, 4.6 x 100 mm, Stainless Steel
Supplier:  NANOMICRO TECHNOLOGIES INC A06003001204610S
Encompass_Preferred
ChromCore Polar C18 columns are based on amide-embedded C18 modified monodispersed porous silica particles with exhaustive end-capping designed for applications that require highly aqueous mobile phase and/or selectivity different from conventional C18 columns
Catalog No. 50-003-32533