Learn More
Medchemexpress LLC Chromeazurol S | 1667-99-8 | 5 G
SDP
Supplier:  Medchemexpress LLC HYD08005G
Encompass_Preferred
Mordant Blue 29, also known as Chromeazurol S, is an azo dye used as an indicator in analytical chemistry to detect metal ions like copper, nickel, and cobalt. It changes color from yellow to blue in the presence of metal ions. It can be applied to a test strip or added directly to a solution for analysis. It is also a biochemical reagent for life science related research.
  • Biochemical reagent for life science research.
Catalog No. 50-004-10925