Learn More
Strem, An Ascensus Company CAS# 10060-12-5. 500g. Chromium(III) chloride hexahydrate. MFCD00149660
SDP
Supplier:  Strem, An Ascensus Company 932417500G
Encompass
CAS# 10060-12-5. 500g. Chromium(III) chloride hexahydrate. MFCD00149660. Molecular Weight: 158.35 (266.48). Molecular Formula: CrCl3∙6H2O. Color/form: green xtl. Strem# 93-2417. http://www.strem.com/catalog/v/93-2417/
Catalog No. 50-901-14546