Learn More
Sigma Aldrich Fine Chemicals Biosciences ChromiumIII potassium sulf500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 243361500G
Encompass
Chromium(III) potassium sulfate dodecahydrate (Potassium chromium(III) sulfate dodecahydrate Chrome alum) is widely used chemical in dyeing and textile industry. It can be synthesized by reacting sulfur dioxide with potassium dichromate (dissolved in sulfuric acid) at -38C.
Catalog No. 50-289-9369