Learn More
Bachem Ac-Lys-pNA
SDP
Supplier:  Bachem 4004444.0050
Encompass_Preferred
Ac-Lys-pNA Hydrochloride, 50931-35-6, C14H20N4O4, Mr 308.34, 50mg. Chromogenic substrate for gingipain K.
Catalog No. 50-194-7043