Learn More
Medchemexpress LLC Chromotropic acid, disodium salt, dihydrate | 5808-22-0 | MFCD00150612 | >98.5% | 400.29 g·mol⁻¹ | C10H6Na2O8S2 · 2 H2O | 10G
SDP
Supplier:  Medchemexpress LLC HY12306510G
Encompass_Preferred
Chromotropic acid disodium dihydrate is the disodium salt dihydrate of chromotropic acid supplied as a light-brown solid powder used as a chromogenic reagent for colorimetric detection and quantification of formaldehyde and other analytical applications. The compound is suitable for research and laboratory analytical chemistry and is provided in multiple laboratory pack sizes.
  • Chromogenic reagent for formaldehyde detection.
  • Suitable for colorimetric and spectrophotometric assays.
  • Available in multiple pack sizes, including 10 g.
  • Solid powder, light brown to brown in appearance.
  • High assay purity (≥98.5% on anhydrous substance) suitable for analytical use.
  • Provided with safety data sheet and product documentation.
Catalog No. 50-002-38793