Learn More
Ichrom Solutions CHROSPE C18, 7 UM CARTRIDGE R
SDP
Supplier:  Ichrom Solutions ASSP0621.003
Encompass
 CHROspe C18, 7 um cartridge range: 10 x 1 mm ID, PEEK - 1000 bar cartridges
Catalog No. NC2283928