Learn More
Medchemexpress LLC Chrysanthemic acid 50mg | 10453-89-1 | 168.23 g·mol⁻¹ | C10H16O2 | 50 MG

Supplier: Medchemexpress LLC HY11450250MG
Chrysanthemic acid is a monocarboxylic acid used as a building block and reference material in research on natural and synthetic insecticides. Supplied for research use, it is available as solid powder and as 10 mM DMSO solutions in multiple pack sizes.
- High purity reference compound suitable for insecticide research.
- Chemical formula C10H16O2 and molecular weight 168.23 g·mol⁻¹.
- CAS number 10453-89-1 for unambiguous identification.
- Available in solid and 10 mM DMSO solution formats, including 50 mg packs.
- Provided for research use only; consult the SDS for handling and safety information.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.