Learn More
Medchemexpress LLC Chrysene-d12 | 1719-03-5 | 99.7% | 50 MG

Supplier: Medchemexpress LLC HY121107S50MG
Chrysene-d12 is the deuterium-labeled form of chrysene (C18D12, CAS 1719-03-5) supplied as a high-purity analytical reference standard for research and mass-spectrometry applications.
- Deuterium labeled (d12) tracer for mass spectrometry and isotope-dilution assays.
- High isotopic purity (99.71%) suitable as an internal standard.
- Supplied as a powder; recommended storage at -20°C to maintain stability.
- Used in environmental, toxicology, and analytical applications as a polycyclic aromatic hydrocarbon (PAH) standard.
- Chemical formula C18D12 and molecular weight 240.37 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.