Learn More
Medchemexpress LLC Chryseno[2,3-d]dioxole-2,7(4aH,5H)-dione, decahydro-5-hydroxy-7a-(hydroxymethyl) | 1040085-99-1 | MFCD29920082 | 98.6% | 430.53 g/mol | C25H34O6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1000871MG
Encompass_Preferred
Budesonide impurity C is a characterized synthetic impurity of budesonide supplied as a solid reference material for analytical and impurity-profiling workflows. It is intended for use in method development, validation, and quality control when identifying or quantifying related substances.
  • Intended for analytical reference and impurity profiling.
  • High purity, 98.6% (by HPLC), suitable for reference standards.
  • Defined chemical characteristics: C25H34O6, MW 430.53 g/mol.
  • Available in milligram quantities for laboratory use.
  • Stable as a powder at -20 °C; solvent storage guidance available in SDS.
  • Accompanied by a datasheet and certificate of analysis for batch-specific data.
Catalog No. 50-002-15182