Learn More
Medchemexpress LLC HY-N2376 5mg Medchemexpress, Chrysin-7-O-glucuronide CAS:35775-49-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN23765MG
Encompass_Preferred
Medchemexpress, HY-N2376 5mg Chrysin-7-O-glucuronide CAS:35775-49-6 Chrysin-7-O-glucuronide is a flavonoid extracted from Scutellaria baicalensis, with antioxidant activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3924