Learn More
Medchemexpress LLC HY-121471 1mg Medchemexpress, Chrysoeriol CAS:491-71-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1214711MG
Encompass_Preferred
Medchemexpress, HY-121471 1mg Chrysoeriol CAS:491-71-4 Chrysoeriol, a natural flavonoid extracted from the tropical plant Coronopus didymus, exhibits potent antioxidant activity. Chrysoeriol shows significant inhibition of lipid peroxidation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0967