Learn More
Medchemexpress LLC Chrysoeriol (standard) | 491-71-4 | 99.9% | 1 MG

Supplier: Medchemexpress LLC HY121471R1MG
Chrysoeriol is an analytical reference standard (CAS 491-71-4) of the flavone chrysoeriol supplied as a 1 mg solid for research and analytical applications including assay calibration, method development, and identity confirmation. Reported purity is 99.89% and molecular weight is 300.26 g/mol.
- Analytical reference standard for assay calibration and method development.
- High reported purity of 99.9% for quantitative applications.
- Chemical formula C16H12O6 and molecular weight 300.26 g/mol.
- Supplied as a 1 mg solid suitable for analytical workflows.
- Appropriate for identity confirmation and quantitative assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.