Learn More
Medchemexpress LLC Chrysosplenetin | 603-56-5 | 374.34 | 20 MG
SDP
Supplier:  Medchemexpress LLC HYN145720MG
Encompass_Preferred
Chrysosplenetin is an orally active polymethoxyflavone that exhibits anticancer effects, acts as an antimalarial sensitizer, and promotes bone formation.
  • Exerts anticancer effects by inhibiting topoisomerase, protecting DNA, and inducing apoptosis.
  • Acts as an antimalarial sensitizer, reversing Artemisinin resistance by inhibiting and downregulating P-glycoprotein, and enhancing efficacy.
  • Promotes bone formation by activating the Wnt/β-catenin pathway, demonstrating anti-osteoporotic effects.
Catalog No. 50-004-14561