Learn More
Medchemexpress LLC Chst15-IN-1 | 2158198-77-5 | 98.0% | 442.09 | C17H11BrCl2N2O3 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1247045MG
Encompass_Preferred
Chst15-IN-1 is a potent, reversible covalent inhibitor of the Chst15 sulfotransferase that reduces chondroitin sulfate-E (CS-E) sulfation and inhibits related glycosaminoglycan (GAG) sulfotransferases. It can diminish the inhibitory effects of chondroitin sulfate proteoglycans (CSPGs) and has been used in research to promote neuronal repair.
  • Potent reversible covalent inhibition of Chst15.
  • Reduces CS-E sulfation and inhibits related GAG sulfotransferases.
  • Diminishes CSPG-mediated inhibition to support neuronal repair.
  • Available as high-purity solid quantities and solution formulations.
  • Well characterized for molecular weight and formula for analytical use.
Catalog No. 50-002-36368