Learn More
Carbosynth LLC. Chymostatin, 25mg, Mixture of Type A, B, and C[(S)-1-Carboxy-2-phenylethyl]-carbamoyl-a-[2-iminohexahydro-4(S)-pyrimidyl]-(S)-glycyl-X-phenylalaninal X: L-leucyl (Type A), L-valyl (Type B), L-isoleucyl (Type C), MW: 610.72, Molecular formula: C31H42N6O7, CAS# 9076-44-2
SDP
Supplier:  Carbosynth LLC. ICY406325MG
Encompass_Preferred
Synonyms: [(S)-1-Carboxy-2-phenylethyl]carbamoyl-a-[2-iminohexahydro-4(S)-pyrimidyl]-(S)-glycyl-X-phenylalaninal, Application: Microbial Product Inhibitor for Chymotrypsin, Papain, and Cathepsin B/G, Storage Temperature: -20°C, References: H. Umezawa, T. Aoyagi, H. Morishima, S. Kunimoto, M. Matsuzaki, M. Hamada, and T. Takeuchi, J. Antibiotics, 23, 425 (1970). (Original), K. Tatsuta, N. Mikami, K. Fujimoto, S. Umezawa, H. Umezawa, and T. Aoyagi, J. Antibiotics, 26, 625 (1973). (Chem. Structure), L.A. Johnson, K.E. Moon, and M. Eisenberg, Biochim. Biophys. Acta, 953, 269 (1988). (Inhibitory Activity)
Catalog No. 50-168-5845